Learn More
Bon Opus Biosciences Recombinant Human AUGER/RAGE (C-6His) - Source: Human Cells
SDP
Supplier:  Bon Opus Biosciences BP30350UG
Encompass_Preferred
Recombinant Human AGER is produced by our Mammalian expression system and the target gene encoding Ala23-Ala344 is expressed with a 6His tag at the C-terminus. Accession#Q15109. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4.
Catalog No. NC2733767