Learn More
Bon Opus Biosciences Recombinant Human BCMA (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW1350UG
Encompass_Preferred
Recombinant Human Tumor Necrosis Factor Receptor Superfamily Member 17 is produced by our Mammalian expression system and the target gene encoding Met1-Ala54 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# Q02223. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CW13
Catalog No. 50-208-0410