Learn More
Lonza Walkersville Hu CB CD34 Cells 500k
Supplier:  Lonza Walkersville 2C101A
Encompass
Cord blood (commonly abbreviated HUCB) CD34 cells are known to contain hematopoietic stem and progenitor cells and are ideal for studies relating to transplantation and stem cells as cord blood derived cells tend to be more na ve than similar cells derived from other tissue sources High Purity and ViabilityLonza Cord Blood CD34 Cells are isolated from cord blood mononuclear cells (MNCs) via positive immunomagnetic separation and are guaranteed to have a purity of 90% as indicated by CD34 expressing cells They are guaranteed to contain 500 000 viable cells HIV-1 and hepatitis B are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Supported by an Optimized Media SystemHPGM Hematopoietic Growth Medium is recommended for optimal performance in serum-free culturing (additional cytokines may be necessary depending on application) Alternative media containing serum may also be acceptable
Catalog No. 50-004-96376