Learn More
Bon Opus Biosciences Recombinant Human CXCL13 (N-hFc) - Source: Human Cells
SDP
Supplier:  Bon Opus Biosciences BP30650UG
Encompass_Preferred
Recombinant Human CXCL13 (N-hFc) is produced by Human Cells. The target gene encoding Val23-Pro109 is expressed with a N-hFc tag. Accession#O43927. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4.
Catalog No. NC2733777