Learn More
Bon Opus Biosciences Recombinant Human Cytochrome b5 B/CYB5B (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CJ361MG
Encompass_Preferred
Recombinant Human Cytochrome b5 B is produced by our Mammalian expression system and the target gene encoding Lys12-Cys118 is expressed with a 6His tag at the C-terminus. Accession# O43169. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CJ36
Catalog No. 50-217-2811