Learn More
Bon Opus Biosciences Recombinant Human FGF2 (Fibroblast Growth Factor 2) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP07013100UG
Encompass_Preferred
Recombinant Human FGF2 (Fibroblast Growth Factor 2) is produced by E. coli. It is a single, nonglycosylated,polypeptide chain containing 155 amino acids. The sequence of the first six N-terminal amino acids was determined to be MPALPE. Accession#P09038. Formulation: The protein was lyophilized from a concentrated (0.5 mg/ml) solution of PBS.
Catalog No. NC2733618