Learn More
Bon Opus Biosciences Recombinant Human Granulocyte Colony-Stimulating Factor/G-CSF 500ug
SDP
Supplier:  Bon Opus Biosciences C002500UG
Encompass_Preferred
Recombinant Human Granulocyte Colony-Stimulating Factor is produced by our E.coli expression system and the target gene encoding Thr31-Pro204 is expressed. Accession# P09919-2. Formulation: Lyophilized from a 0.2 μm filtered solution of 10mM HAc-NaAc, 150mM NaCl, 0.004% Tween 80, 5% Mannitol, pH 4.0. Bon Opus Cat# C002
Catalog No. 50-217-2715
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.