Learn More
Bon Opus Biosciences Recombinant Human Isocitrate Dehydrogenase 1/IDH1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C9431MG
Encompass_Preferred
Recombinant Human Isocitrate Dehydrogenase is produced by our Mammalian expression system and the target gene encoding Met1-Leu414 is expressed with a 6His tag at the C-terminus. Accession# O75874. Formulation: Supplied as a 0.2 μm filtered solution of PBS. Bon Opus Cat# C943
Catalog No. 50-217-2765