Learn More
Bon Opus Biosciences Recombinant Human Interferon Regulatory Factor 5/IRF5 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CF181MG
Encompass_Preferred
Recombinant Human Interferon Regulatory Factor 5 is produced by our E.coli expression system and the target gene encoding Met1-Gln498 is expressed with a 6His tag at the C-terminus. Accession# Q13568. Formulation: Supplied as a 0.2 μm filtered solution of 20mM Tris-HCl, 150mM NaCl, 30% Glycerol, 1mM DTT, pH8.0. Bon Opus Cat# CF18
Catalog No. 50-217-2783