Learn More
Bon Opus Biosciences Recombinant Human Lymphocyte Activation Gene 3/LAG-3/CD223 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CJ911MG
Encompass_Preferred
Recombinant Human LAG-3 is produced by our Mammalian expression system and the target gene encoding Leu23-Gly434 is expressed with a 6His tag at the C-terminus. Accession# P18627. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CJ91
Catalog No. 50-217-2816