Learn More
Cell Biologics Human Primary Vein Endothelial Cells, 1 Frozen Vial
SDP
Supplier:  Cell Biologics H6009
Encompass_Preferred
Human Primary Vein Endothelial Cells from Cell Biologics are isolated from human vein tissue and grown in gelatin pre-coated tissue culture flask with Cell Biologics’ Complete Growth Medium. Cells at passage 3 are harvested from flasks and cryo-preserved in vials. Each vial contains 0.5x106 cells per ml and is delivered frozen. Human Primary Vein Endothelial Cells are characterized by immunofluorescence staining with antibodies of CD31 (Catalog No. 550389, BD), CD31/PECAM-1 (Catalog No. FAB3567P, R&D), VE-Cadherin (FITC-VE-cadherin, Catalog No. 560411, BD), or use of fluorescence-labeled acetylated low-density lipoprotein (DiI-Ac-LDL) uptake (Catalog No. L-35353, Invitrogen), a functional marker for endothelial cells. These cells are negative for bacteria, yeast, fungi, and mycoplasma and can be expanded for 3-5 passages at a split ratio of 1:2 under the cell culture conditions specified by Cell Biologics. Repeated freezing and thawing
Catalog No. NC2736560