Learn More
Bon Opus Biosciences Recombinant Human Disks Large Homolog 4/DLG4/PSD95 (N-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CH23500UG
Encompass_Preferred
Recombinant Human Postsynaptic Density Protein 95 is produced by our E.coli expression system and the target gene encoding Met1-Leu724 is expressed with a 6His tag at the N-terminus. Accession# P78352. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CH23
Catalog No. 50-217-2793