Learn More
Bon Opus Biosciences Recombinant Human Sentrin-Specific Protease 7/SENP7 (N-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C171500UG
Encompass_Preferred
Recombinant Human Sentrin-Specific Protease 7 is produced by our E.coli expression system and the target gene encoding Met695-Ala864 is expressed with a 6His tag at the N-terminus. Accession# Q9BQF6. Formulation: Supplied as a 0.2 μm filtered solution of 20mM HEPES, 5% Glycerol, pH 7.4. Bon Opus Cat# C171
Catalog No. 50-217-2726