Learn More
Bon Opus Biosciences Recombinant Human Serpin B12/SERPINB12 (C-6His) 1mg
Supplier: Bon Opus Biosciences CI961MG
Recombinant Human Serpin B12 is produced by our Mammalian expression system and the target gene encoding Met1-Pro425 is expressed with a 6His tag at the C-terminus. Accession# Q96P63. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CI96