Learn More
iQ Biosciences Human Treg Suppression Assay Kit w/ CFSE-Labeled CD8 T Cells - 5 pk
Supplier:  iQ Biosciences IQBKA0025C
Encompass
Human Treg functional kits with Regulatory T cells and CD8 T cell responders. CD8 T cells are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis
Catalog No. NC2855554
Only null left
Add to Cart
Request a Quote