Learn More
Bon Opus Biosciences Recombinant Human Vinculin 500ug
SDP
Supplier:  Bon Opus Biosciences C264500UG
Encompass_Preferred
Recombinant Human Vinculin is produced by our E.coli expression system and the target gene encoding Pro2-Gln1066 is expressed. Accession# P18206. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C264
Catalog No. 50-217-2731