Learn More
Bon Opus Biosciences Recombinant Human Zinc Finger MYND Domain-Containing Protein 19/ZMYND19 (N-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences C280500UG
Encompass_Preferred
Recombinant Human Zinc Finger MYND Domain-Containing Protein 19 is produced by our E.coli expression system and the target gene encoding Met1-Arg227 is expressed with a 6His tag at the N-terminus. Accession# Q96E35. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C280
Catalog No. 50-217-2733