Learn More
Bon Opus Biosciences Recombinant Human Haptoglobin/Zonulin (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP14350UG
Encompass_Preferred
Recombinant Human Haptoglobin is produced by Insect Cells and the target gene encoding V19-N406 is expressed with a 8His tag at the C‐terminus. Expression system: Insect Cells. Accession# P00738. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP143
Catalog No. 50-208-0227