Learn More
Cayman Chemical HU331 Analytical Reference Sta
SDP
Supplier:  Cayman Chemical 1000567310 MG
Encompass_Preferred
An analytical reference standard categorized as an oxidative degradation product of CBD; intended for research and forensic applications
Catalog No. 50-288-8926