Learn More
iQ Biosciences Huacaya Alpaca PBMCs, 50M cells/vial
Supplier:  iQ Biosciences IQBAPPB150
Encompass
High-quality, high-viability (> 80%) huacaya alpaca peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Potential uses include, production of stable single-domain antibodies (nanobodies) for therapeutic and diagnostic applications, vaccine development, and autoimmune research. All orders come with an iQ Certificate of Analysis.
Catalog No. NC3235561