Learn More
Promocell GmbH PromoCell HUASMC-c 500,000 Human Umbilical Artery Smooth Muscle Cells, cryopreserved
Supplier:  Promocell GmbH C12500
Encompass
Primary Human Umbilical Artery Smooth Muscle Cells (HUASMC) are isolated from the arteries of the umbilical cord and stain positive for smooth muscle α-actin. Physiological and biochemical processes of vascular diseases can be analyzed with HUASMC in culture.
Catalog No. 50-306-024