Learn More
MSC Hubbell Workplace Solutions Air Torque Stabilizer, 0.5-1.5 Lb, Rail Mount
Supplier:  MSC 78810827
Encompass
This air torque stabilizer provides ergonomic support for air or electric powered tools weighing between 0.5 and 1.5 pounds. It features all-steel construction with a baked polyester finish, a swing boom, and a trolley system for precise tool positioning.
  • All steel construction with a durable baked polyester finish.
  • Swing boom with trolley for precise tool positioning.
  • Boom, bracket, and swing arms are height adjustable.
  • U-joint manifold supplies air and allows vertical and horizontal tool positioning.
  • Includes a tool balancer and telescoping positioning arms.
  • Ergonomic design prevents torque transmission to the worker.
  • Aids OSHA ergonomics compliance.
  • Mounting base has four pre-drilled 0.56 inch diameter holes.
Catalog No. 50-005-30784