Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human 4-1BB (TNFRSF9) Protein, His Tag, MALS-verified - 100 μg

Supplier:  ACROBiosystems 41BH53H3100UG

Encompass_Preferred

Recombinant human 4-1BB (TNFRSF9) extracellular domain expressed in CHO cells, supplied with a C-terminal His tag, and verified by SEC-MALS for monodispersity and molecular weight. Suitable for binding studies, antibody screening, ELISA, and assay development.

  • Expressed in CHO cells for mammalian-type glycosylation.
  • Contains residues Leu 24-Gln 186 of human 4-1BB.
  • C-terminal polyhistidine tag for purification and detection.
  • Purity greater than 90% by SDS-PAGE and greater than 95% by SEC-MALS.
  • Molecular weight approximately 22-32 kDa as measured by SEC-MALS.
  • Available in 100 μg and 1 mg sizes for assay flexibility.
  • Validated for in vitro binding assays, antibody characterization, and assay controls.

Catalog No. NC2744407


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.