Learn More
Cellular Engineering Technologies Cryopreserved vial of induced pluripotent stem cells from a single male donor with Cystic Fibrosis (500,000 cells)
Supplier:  Cellular Engineering Technologies CR1010500
Encompass_Preferred
Cell line was derived from a single caucasian male donor diagnosed with Cystic Fibrosis.Cystic fibrosis is caused by mutations in the gene encoding the cystic fibrosis transmembrane conductance regulator (CFTR) protein, which is involved in mucus production. This genetic disorder primarily affects the lungs, pancreas, liver, kidneys, and intestines.Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753015