Learn More
Cell Biologics Human Endothelial Cell Medium (Glucose and Phenol Red Free) – 500 ML
SDP
Supplier:  Cell Biologics H1168GPF
Encompass_Preferred
Endothelial Cell Medium (Glucose and Phenol Red Free) is a medium designed for the culture of human and Monkey endothelial cells. It was tested and optimized with cell growth and proliferation in vitro. It is formulated for use with 5% CO2 and 95% air in a humidified incubator. The medium consists of 500 ml of basal medium (containing essential and non-essential amino acids, vitamins, organic and inorganic compounds, hormones, growth factors, trace minerals).
Catalog No. NC2960383