Learn More
ACROBiosystems Recombinant Protein;Human Fc gamma RIIB / CD32b Protein (SPR verified);HEK293;100UG;CDB,FCGR2B,CD32b,CD32,FCG2,IGFR2,CDw32,FCG-2,IGFR-2,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems CDBH5228100UG
Encompass_Preferred
Recombinant Protein;Human Fc gamma RIIB / CD32b Protein (SPR verified);100UG;Human CD32b (SPR verified) (CDB-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession # P31994).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.4 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CD32b (Cat. No. CDB-H5228) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 10.1 μM as determined in a SPR assay (Biacore T200) (QC tested). ;Immobilized Rituximab on CM5 Chip, can bind Human CD32b (Cat. No. CDB-H5228) with an affinity constant of 5.5 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-101-7475