Learn More
Cellular Engineering Technologies Keratinocyte Growth Media (450mL)
Supplier:  Cellular Engineering Technologies MR1013
Encompass_Preferred
Human Keratinocyte Growth Media is designed to support robust growth keratinocyte proliferation. When using this media, keratinocyte cells have been observed to retain cuboidal morphology and a 48-hour doubling time. Media is shipped with gel packs.
Catalog No. NC2753086