Learn More
IPHASE PHARMA SERVICES LLC Human MATE2-K SLC Transporter Cells,8-10million
Supplier: IPHASE PHARMA SERVICES LLC 01943A1.01
Participate in drug-drug interactions and drug uptake/ drug efflux. SLC transporter developed using transfected cells as a method and HEK293F cells as a vector.They play key roles in drug resistance, metabolism, and cellular detoxification and usually be used in DDI studies. The product need to be used in conjunction with MOCK/HEK293F Cell, Transporter Buffer and Polylysine Coated Plate.These products require shipment in LN2 container therefore customer need to return the container.