Learn More
IPHASE PHARMA SERVICES LLC Human OAT3 SLC Transporter Cells,8-10million
Supplier:  IPHASE PHARMA SERVICES LLC 01945A1.01
Encompass_Preferred
Participate in drug-drug interactions and drug uptake/ drug efflux. SLC transporter developed using transfected cells as a method and HEK293F cells as a vector.They play key roles in drug resistance, metabolism, and cellular detoxification and usually be used in DDI studies. The product need to be used in conjunction with MOCK/HEK293F Cell, Transporter Buffer and Polylysine Coated Plate.These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3966952