Learn More
Abcam Human TGF beta 1 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1006471X96TESTS
Encompass_Preferred
Human TGF beta 1 ELISA kit (colorimetric) is a sandwich ELISA designed to quantify transforming growth factor beta 1 (TGF-β 1) with a sensitivity of 18 pg/mL. - Wide dynamic range – quantifies 18–4,000 pg/mL - Broad sample compatibility – works with serum, plasma, and cell culture supernatants - Cited in over 85 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2077855