Learn More
Biotang Inc Human urokinase plasminogen activator,uPA ELISA Kit, 96T
Supplier: Biotang Inc HU8886
Human urokinase plasminogen activator,uPA ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.