Learn More
Sino Biological Human 2B4/CD244 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK100425
Encompass_Preferred
The Human 2B4 / CD244 ELISA Pair Set is for the quantitative determination of Human 2B4 / CD244.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for 2B4 / CD244 coated on a 96-well plate. Standards and samples are added to the wells, and any 2B4 / CD244 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-2B4 / CD244 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of 2B4 / CD244 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86621