Learn More
ACROBiosystems ACROBiosystems Human ACE2 / ACEH Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems AC2H52H4200UG
Encompass_Preferred
Recombinant Protein;200UG;Human ACE2, His Tag (AC2-H52H4) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 85.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H4) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein RBD, Fc Tag (Cat. No. SPD-C5255) with a linear range of 0.1-0.8 ng/mL (QC tested).;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H4) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1N-C5255) with a linear range of 0.4-6 ng/mL (Routinely tested).
Catalog No. 50-210-7427