Learn More
American Research Products Inc Human actiVated coagulation fa
Supplier:  American Research Products Inc CSBE12696H
Encompass
Human actiVated coagulation factor X (FXa) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29147