Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Human Adenovirus 2 (ATCC VR-1786), strain ATCC-2011-7 - frozen BSL-2 research reference

Supplier:  ATCC VR1786

Encompass

Human adenovirus 2 is an authenticated, genome-sequenced viral reference strain supplied for laboratory research applications such as virology studies, molecular characterization, and assay development. This material is provided for research use only and is not intended for therapeutic, diagnostic, or consumption purposes.

  • Authenticated reference strain suitable for research use.
  • Genome-sequenced with a reported length of approximately 35.7 Kb.
  • Classification: human adenovirus C (Mastadenovirus genus).
  • Provided in a frozen format for integrity preservation.
  • Storage recommendation: maintain at -70°C or colder.
  • Biosafety level: BSL-2; handle according to institutional biosafety guidelines.
  • Appropriate for assay development, molecular studies, and virology research.

Catalog No. NC2876620


May include imposed supplier surcharges.

Due to product restrictions, please to purchase or view availability for this product.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.