Learn More
ATCC Human adenovirus 2; Strain: Adenoid 6
Supplier:  ATCC VR846
Encompass
Human adenovirus 2 strain Adenoid 6 is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated in 1953 from a 7-year-old human female with hypertrophied tonsils and adenoids and was deposited by NIH/NIAID. This product has applications in infectious and respiratory research.
Catalog No. 50-238-2792
 Restricted Item