Learn More
ATCC Human adenovirus 3; Strain: G.B.
Supplier:  ATCC VR3
Encompass
Human adenovirus 3 strain GB is propagated in A549 cells (ATCC CCL-185). This strain was isolated in 1953 from the nasal washings of an adult male with a common cold and was deposited by National Institute of Allergy and Infectious Diseases. It has applications in respiratory disease research.
Catalog No. 50-238-1917