Learn More
ATCC Human adenovirus 4; Strain: RI-67
Supplier:  ATCC VR1572
Encompass
Human adenovirus 4 strain RI-67 is propagated in HeLa 229 cells (ATCC CCL-2.1). This strain was isolated in 1953 from the throat washings of a patient at Fort Leonard Wood in Missouri, Kansas, and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.
Catalog No. 50-238-2957