Learn More
ATCC Human adenovirus 41; Quantitative DNA
Supplier:  ATCC VR930DQ
Encompass
Quantitative Genomic DNA from Human adenovirus 41 strain Tak (73-3544) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2391