Learn More
ATCC Human adenovirus 7a (ATCC VR-848), strain S-1058
Supplier: ATCC VR848
Human adenovirus 7a (strain S-1058) is a characterized viral isolate used for infectious and respiratory disease research. The virus produces a cytopathic effect in permissive cell lines and is reported to be weakly oncogenic; propagation and storage conditions are provided for laboratory use.
- Strain designation: S-1058.
- Organism: Human adenovirus 7a.
- Taxonomy: Mastadenovirus (Human mastadenovirus B).
- Observed effects: causes cytopathic effect and is weakly oncogenic.
- Storage: -70°C or colder.
- Propagation medium: EMEM with 2% fetal bovine serum.
- Incubation: 2 to 3 days at 37°C.
- Atmosphere: 95% air, 5% CO2.
- Infection recommendation: infect cells that are 24 hours old and 70%-80% confluent for best results.
- Depositor: NIH/NIAID.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.