Learn More
Abcam Human Adiponectin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB999681X96TESTS
Encompass_Preferred
Human Adiponectin ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Adiponectin in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2330689