Learn More
Sino Biological Human AK4 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12406MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AK4 / AK3L1 (rh AK4 / AK3L1; Catalog#12406-H20B; P27144-1; Ala 2-Tyr 223). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1038