Learn More
Rockland Immunochemicals Human Albumin Biotin Conjugated 1 mg
SDP
Supplier:  Rockland Immunochemicals 0090633
Encompass
Human albumin or serum albumin is encoded by the ALB gene and is the most abundant plasma protein in mammals. Human albumin is essential for maintaining the osmotic pressure needed for proper distribution of body fluids between intravascular compartments and body tissues. Human albumin also acts as a plasma carrier by non-specifically binding several hydrophobic steroid hormones and as a transport protein for hemin and fatty acids. Too much serum albumin in the body can be harmful.
Catalog No. 50-194-3757