Learn More
ACROBiosystems Human Alpha-Synuclein (A53T) Protein, Tag Free (MALS verified)
Supplier:  ACROBiosystems ALNH5116200UG
Encompass_Preferred
Human Alpha-Synuclein (A53T), Tag Free (ALN-H5116) is expressed from E. coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1 (A53T)).
Catalog No. 50-233-1899