Learn More
Enzyme Research Laboratories Inc Human a-Thrombin (Factor IIa),1000 U
Supplier:  Enzyme Research Laboratories Inc HT 1002A10 OR MORE
Encompass
Please use this part # if ordering a quantity of 10 or more. If less than qty 1-9 please use Fisher part # NC9872266.
Human a-Thrombin (Factor IIa),1000 U,Buffer Composition = 50 mM Sodium Citrate,0.2 M NaCl,0.1% PEG-8000,pH 6.5,Extinction Coefficient (1%) = 18.3,Molecular Weight = 37,000 daltons,Prepared from human Prothrombin by activation with Factor Xa
Catalog No. NC0704726