Learn More
Abcam Human ALT ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2345781X96TESTS
Encompass_Preferred
Human ALT ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human alanine aminotransferase (ALT) with a sensitivity of 47.34 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA with a 450 nm readout; compatible with any standard plate reader - Different formats for different needs, including 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready carrier-free antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1634504