Learn More
Raybiotech Inc C-Series Human Angiogenesis Antibody Array 2 Kit.
Supplier:  Raybiotech Inc AAHANG22
Encompass_Preferred
Detects 23 Human Angiogenic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Angiopoietin-1, Angiopoietin-2, Angiostatin, Endostatin, GCSF, GM-CSF, I-309 (TCA-3/CCL1), IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, I-TAC (CXCL11), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MMP-1, MMP-9, PECAM-1 (CD31), Tie-2, TNF alpha, uPAR, VEGFR2, VEGFR3
Catalog No. 50-194-8468