Learn More
ACROBiosystems Human Angiopoietin-like 4 / ANGPTL4 (166-406) Protein, His Tag
Supplier: ACROBiosystems AN4H5243100UG
Recombinant Protein;100UG;Human Angiopoietin-like 4 (166-406), His Tag (AN4-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession # Q9BY76-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 33-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.