Learn More
Cytiva Human Antibody Capture Kit, type 2
Supplier:  Cytiva 29234600
Encompass
Reagents for capture of human or humanized IgG antibodies as ligands in biomolecular interaction analyses. Designed for use with Biacore 8K (larger kit component volumes than in Human Antibody Capture Kit) but can be used with all other Biacore systems.
Catalog No. 50-152-8214