Learn More
American Research Products Inc Human Antigen KI 67 Ki 67 EL
Supplier:  American Research Products Inc CSBE16294H
Encompass
Human Antigen KI-67 (Ki-67) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28595